Glass-Filled Polypropylene Tube Connectors, Ace Glass Incorporated
# de catalogue 89077-386
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Elbow Tube to Tube
- Cat. no.:89077-386
- Fits Tubing O.D.:12.7 mm (½")
- Supplier No.:12705-17
Spécifications
A propos de cet article
These glass-filled polypropylene connectors are designed for use with stainless steel, nylon, polyolefin, aluminum, and all other tubing.
Customers Who Bought This Also Bought