ContactUsLinkComponent
12705 Glass-filled polypropylene elbow tube to tube.
Glass-Filled Polypropylene Tube Connectors, Ace Glass Incorporated
# de catalogue 89077-386
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
12705 Glass-filled polypropylene elbow tube to tube.
Glass-Filled Polypropylene Tube Connectors, Ace Glass Incorporated
# de catalogue 89077-386
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    Elbow Tube to Tube
  • Cat. no.:
    89077-386
  • Fits Tubing O.D.:
    12.7 mm (½")
  • Supplier No.:
    12705-17
Spécifications
A propos de cet article
These glass-filled polypropylene connectors are designed for use with stainless steel, nylon, polyolefin, aluminum, and all other tubing.
Customers Who Bought This Also Bought