Pluro Stopper Set, Ace Glass Incorporated
# de catalogue 89075-804
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Pluro Stopper Set
- Cat. no.:89075-804
- For O.D.:18-70 mm
- Supplier No.:12014-14
Spécifications
A propos de cet article
Versatile silicone stopper set that equals 17 standard stoppers
All the rings are cut from the same stopper, with each ring nesting perfectly into the next. Whether making up a small or large stopper, a vacuum-tight fit is assured. Sold as a set.
Customers Who Bought This Also Bought