Micro Reaction Vials, Conical Bottom, Ace Glass Incorporated
# de catalogue 89075-240
Fournisseur: Ace Glass
Small Business Enterprise
Spécifications
- Cap included:Yes
- Cap Size:15-415
- Capacity:5.00 ml
- Description:Micro reaction vials, 10/10 outer joint
- Cat. no.:89075-240
- Bottom Style:Conical
- Supplier No.:9590-19
Spécifications
A propos de cet article
Microscale, heavy-wall reaction vial.
The top joint is a combination cap, thread and joint to accommodate and seal other microscale apparatus. The Ext.Ø. of the top joint has an external thread to facilitate an O-ring and open-top phenolic cap. The internal or Int.Ø. at the top is a ground outer joint. This type connection eliminates the need for clamps while offering the positive leaktight seal of ground glass joints.
Replace O-ring in cap with PTFE-faced silicone rubber septa and vial can be capped for storage.
Delivery: Each vial supplied with threaded cap with hole, one PTFE faced silicone rubber septum and one O-ring for making 'O-Ring-CAP-SEAL.'
Customers Who Bought This Also Bought