Button Type Glass Stirring Shaft, 5 and 6 mm, Ace Glass Incorporated
Catalog # 89075-082
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Polished Glass Stirring Shaft
- Cat. no.:89075-082
- Shaft Diameter:6 mm
- Supplier No.:9534-40
Specifications
About this item
There is a button on a 5 and 6 mm outer Ø stir rod for the ground glass stirring shaft. This stirring shaft has a total length of 318 mm.
Customers Who Bought This Also Bought