WorkWipes® Series 60 Heavy-Duty Wipers, New Pig
# de catalogue 89073-946
Fournisseur: 2 D 2 S.A. DE C.V. MX
Spécifications
- W×L:30.5x30.5 cm (12x12")
- Cat. no.:89073-946
- Packaging:Jumbo roll (1100 sheets)
- Weight:6.7 kg (14.8 lbs.)
- Supplier No.:WIP661
Spécifications
A propos de cet article
Lightest of the heavy-duty WorkWipes® series, these economical wipers are made of cellulose and synthetic fibers for strength and low linting properties.
- Textured surface helps remove dirt and grime
- Ideal for applying disinfectants, degreasers, and solvents
Additional applications include grease removal, wiping hands, fast cleanups, and cleaning glass or stainless steel.
Customers Who Bought This Also Bought