ContactUsLinkComponent
VWR* Hi-Den Liner 30X37X 13 Mic
VWR® High Density Liner Bags
# de catalogue 89072-212
Fournisseur:  VWR International
VWR* Hi-Den Liner 30X37X 13 Mic
VWR® High Density Liner Bags
# de catalogue 89072-212
Fournisseur:  VWR International
Spécifications
  • W×L:
    762×940 mm (30×37")
  • Thickness:
    13 µm
  • Cat. no.:
    89072-212
Spécifications
A propos de cet article
High Density Polyethylene (HDPE) liners provide extra strength at lighter weight.
  • Provided in heavy-duty dispenser box
  • Available in a range of thicknesses
High Density Polyethylene (HDPE) liners provide extra strength at lighter weight.
Provides excellent puncture resistance from sharp objects and heavy items. Comes in convenient heavy-duty dispenser box. Star sealed bottoms help prevent tear-out.
Customers Who Bought This Also Bought