Order Entry
Canada
ContactUsLinkComponent
VWR® Recloseable Bags with Sliding Closure
Catalogue # 89071-924
Fournisseur:  VWR International
undefined
VWR® Recloseable Bags with Sliding Closure
Catalogue # 89071-924
Fournisseur:  VWR International

Spécifications

  • Thickness:
    3 mil
  • Cat. no.:
    89071-924
  • W×L:
    229×305 mm (9×12")

Spécifications

À propos de cet article

Reclosable bags are full gauge and offer exceptional clarity and quality.

Slider top bag opening is approximately 4.4 cm (1³/₄") smaller than specified bag width to accommodate the slider closure.

Certifications: FDA approved.
Frequently Bought Together