Spécifications
- Color:Clear
- Description:Triangle-Shaped Restraints
- Size:Large
- Cat. no.:89066-362
- Supplier No.:1803C
Spécifications
A propos de cet article
Rat/mouse restraints are ideal for tail tattooing, tail vein injections, lp injections, oral dosing, decapitation, and general restraint
Humane, safe, and disposable. Cone-shaped bags provide easier animal capture and increased animal comfort.