ContactUsLinkComponent
Permanent Green Tattoo Pigment
Animal Tattoo Pigments, Needles, and Supplies, AIMS™
# de catalogue 89066-288
Fournisseur:  Aims
Minority or Woman-Owned Business Enterprise
Permanent Green Tattoo Pigment
Animal Tattoo Pigments, Needles, and Supplies, AIMS™
# de catalogue 89066-288
Fournisseur:  Aims
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Permanent Green Tattoo Pigment
  • Cat. no.:
    89066-288
  • Supplier No.:
    2700
Spécifications
A propos de cet article
For use with AIMS™ animal tattoo systems (see 89066-278 series).
Needles, made from 401 stainless steel and silver solder, are provided presterilized and ready for tattooing. Needle design minimizes animal pain and tissue damage.
Pigments have been demonstrated to be safe and nontoxic using in vivo and in vitro toxicology assays. They will not alter the physiology of animals or contaminate other lab animals in the facility.
Customers Who Bought This Also Bought