Septa, Sleeve Type, Rubber, Ace Glass Incorporated
Catalogue # 89064-924
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Color:Red
- Description:For [ST]24/40, [ST]24/25 Joints
- Cat. no.:89064-924
- Supplier No.:9096-156
Spécifications
À propos de cet article
This sleeve-type septa has a hollow plug
The top is flanged with a sleeve-like extension that folds down over the neck of vessel. The diaphragm can be punctured readily with a syringe needle. Puncture seals automatically after the needle is withdrawn.
Ordering information: Rubber septum are available in white, gray, or red finishes.
Frequently Bought Together