O-Ring Replacement Sets for Stopcocks, Ace Glass Incorporated
# de catalogue 89062-462
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Kalrez O-Rings
- Stopcock Size:0 - 15 mm
- Supplier No.:8194-130
Spécifications
A propos de cet article
Replacement O-Ring sets are designed for use with stopcock plugs.
Available in five different compounds.
Packaging: Packaging includes two large O-Rings and one small O-Ring for the plug sizes specified.
Customers Who Bought This Also Bought