ContactUsLinkComponent
8194 STOPCOCK, High Vacuum, Easy Action Plug
O-Ring Replacement Sets for Stopcocks, Ace Glass Incorporated
# de catalogue 89062-462
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
8194 STOPCOCK, High Vacuum, Easy Action Plug
O-Ring Replacement Sets for Stopcocks, Ace Glass Incorporated
# de catalogue 89062-462
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    Kalrez O-Rings
  • Stopcock Size:
    0 - 15 mm
  • Supplier No.:
    8194-130
Spécifications
A propos de cet article
Replacement O-Ring sets are designed for use with stopcock plugs.
Available in five different compounds.
Packaging: Packaging includes two large O-Rings and one small O-Ring for the plug sizes specified.
Customers Who Bought This Also Bought