ContactUsLinkComponent
7891 TURNTABLE REACTOR, Photochemical
Photochemical Turntable Reactor, Ace Glass Incorporated
# de catalogue 89061-404
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
7891 TURNTABLE REACTOR, Photochemical
Photochemical Turntable Reactor, Ace Glass Incorporated
# de catalogue 89061-404
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    Photochemical Turntable Reactor, with 1/29 HP Motor
  • Cat. no.:
    89061-404
  • Electrical:
    120 V, 50/60 Hz
  • Supplier No.:
    7891-30
Spécifications
A propos de cet article
ACE photochemical reactor with sample roundtable feature for the determination of relative and absolute quantum yields
Polished and Colored Filter Glasses are a combination of either Corning Glass Filter cs 0-52 and Corning Glass Filter cs 7-37 or Corning Glass Filter cs 7-60 isolates 3650 line. 51 x 51 mm. For use in four-sided filter chamber supplied with the Photochemical turntable reactor.
When Corning Glass Filter cs 7-54 is used in conjunction with a filter solution of potassium chromate and sodium hydroxide less than 3% of the 3340 line passes. The solution may be circulated via pump as a coolant through well. 51x51 mm. or use in four-sided filter chamber supplied with the Photochemical turntable reactor.
Features adjustable height slots for up to 33 sample tubes in outer circle and 18 tubes in inner circle, 13 mm O.D. Samples rotate equidistantly around lamp at 6 rpm. Chamber between lamp and samples holds four, 51 mm square, removable flat filters. Unit is constructed of anodized aluminum, brass and PTFE. Measures 25.4 cm diameter at base × 61 cm high at top of motor stand. Except for motor, reactor is completely immersible and can be easily disassembled for cleaning.
Ordering information: Available in 115 V, 50/60 Hz. or 230 V, 50 Hz. Supplied with two-wire, ground NEMA plug. Complete unit includes motor, but does not include filters, immersion well, lamp, power supply or sample tubes.
Customers Who Bought This Also Bought