ContactUsLinkComponent
7855 O-RING
O-Rings, FETFE®, Ace Glass Incorporated
# de catalogue 89061-016
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
7855 O-RING
O-Rings, FETFE®, Ace Glass Incorporated
# de catalogue 89061-016
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    FETFE O-Rings
  • Cat. no.:
    89061-016
  • Supplier No.:
    7855-734
Spécifications
A propos de cet article
FETFE is an exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along with excellent temperature and chemical compatibility. Generally black or white in color.
Customers Who Bought This Also Bought