ContactUsLinkComponent
7759 STORAGE TUBE, Schlenk
Vacuum Filtration Tube, 3 Neck, Ace Glass Incorporated
# de catalogue 89060-388
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
7759 STORAGE TUBE, Schlenk
Vacuum Filtration Tube, 3 Neck, Ace Glass Incorporated
# de catalogue 89060-388
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Capacity:
    250 ml
  • Porosity:
    25-50 µm
  • Description:
    Vacuum Filtration Tube
  • Cat. no.:
    89060-388
  • Joint Size:
    Outer (2) 24/25; Inner (2) 24/40
  • Supplier No.:
    7759-31
Spécifications
A propos de cet article
Use this 3-neck vacuum filtration tube for filtration of solvents and collection of precipitate.
  • Two [ST] outer joints
  • Two [ST] inner joints
  • With fritted disc
This filtration tube has two standard taper 14/20 or standard taper 24/25 outer joints and two standard taper 14/35 or standard taper 24/40 inner. With fritted disc. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought