Borosilicate Glass Spherical Ball Joint with O-Ring Groove, Ace Glass Incorporated
# de catalogue 89060-008
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- O.D.:28
- Cat. no.:89060-008
- ST Joint Size:35/25
- O-Ring Size:−118
- Supplier No.:7649-53
Spécifications
A propos de cet article
Adapter features a spherical ball joint, precision ground, with a groove for an o-ring.
- Leak-proof at high vacuums when clamped
- O-ring permits use without grease
- One FETFE o-ring included
For use with matching socket members. Adapter is leak-proof at high vacuums when clamped. O-Ring permits use without grease. Supplied with one FETFE O-Ring.
Customers Who Bought This Also Bought