Order Entry
ContactUsLinkComponent
7560 EPOXY ADHESIVE
Epoxy Adhesive, Ace Glass Incorporated
# de catalogue 89059-444
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
7560 EPOXY ADHESIVE
Epoxy Adhesive, Ace Glass Incorporated
# de catalogue 89059-444
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise

Spécifications

  • Description:
    Epoxy adhesive, 30 ml
  • Cat. no.:
    89059-444
  • Supplier No.:
    7560-10

Spécifications

A propos de cet article

A two component epoxy system, which cures to a chemical resistant film.

  • Cures to a chemical resistant film
  • Bonds tightly
  • 30mL jar

The epoxy is used to tightly bond a PTFE sleeve to a glass joint. Supplied in 30 mL jars with the activator in a separate container. The 30 mL supply will cement at least 25 joints depending on size.

Customers Who Bought This Also Bought