Gas Dispersion Tubes, Ace Glass Incorporated
# de catalogue 89058-358
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Length:150 mm
- Porosity:C (25 - 50 µm)
- Supplier No.:7202-06
Spécifications
A propos de cet article
Gas dispersion tubes feature a porous fritted glass tip
The O.D. for all sizes is 10mm, and all tubes fit a No. 11 Ace-Thred. Manufactured from borosilicate glass.
Customers Who Bought This Also Bought