Gas Dispersion Tube with Bottom Frit, Ace Glass Incorporated
# de catalogue 89058-312
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Porosity:C (25 - 50 µm)
- Description:Gas Dispersion Tube with Bottom Frit
- Disc Diameter:25 mm
- Supplier No.:7201-06
Spécifications
A propos de cet article
The outside diameter of this gas dispersion tube is 8mm, and the total length of the tube is 150mm
The bottom frit is approximately 2mm thick. Available in two different disc diameters, and three different porosities. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought