Gas Dispersion Tube, Ace Glass Incorporated
# de catalogue 89058-304
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Porosity:D (10 - 20 µm)
- Description:Gas Dispersion Tube
- Disc Diameter:25 mm
- Supplier No.:7196-18
Spécifications
A propos de cet article
This gas dispersion tube's outside diameter is 8mm, and the total length of tube is 150mm
The bottom support is the same O.D. as the disc. Available in two different disc diameters, and five different porosities. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought