ContactUsLinkComponent
6472 Pilot Plant Reactor, Cylindrical, 10L, 30L, 50L
Reaction Head for Pilot Plant, Ace Glass
# de catalogue 89054-654
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
6472 Pilot Plant Reactor, Cylindrical, 10L, 30L, 50L
Reaction Head for Pilot Plant, Ace Glass
# de catalogue 89054-654
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    Reaction head with 7 openings, 300 mm flange
  • Cat. no.:
    89054-654
  • Side ST Joint Size:
    (4)45/50, (2)29/42
  • Supplier No.:
    6472-23
Spécifications
A propos de cet article
Borosilicate glass domed heads with reinforced standard taper joints for pilot plant reactors
Some heads are available with angled addition ports, for ease in adding materials to the reactor.
Heads have either DURAN® or KF flanges, standard taper joint center necks (45/50) and side necks.
Ordering information: Head does not include addition port cap, O-ring, or clamp, they must be ordered separately.
Customers Who Bought This Also Bought