ContactUsLinkComponent
6469 REACTION ASSEMBLY, Portable, 12 and 22L
4" Conical Flange Single Neck Head, 45/50, Ace Glass Incorporated
# de catalogue 89054-502
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
6469 REACTION ASSEMBLY, Portable, 12 and 22L
4" Conical Flange Single Neck Head, 45/50, Ace Glass Incorporated
# de catalogue 89054-502
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    4" Conical Flange Single Neck Head, 45/50
  • Cat. no.:
    89054-502
  • Supplier No.:
    6469-26
Spécifications
A propos de cet article
4" Conical Flange Head with 45/50 Center Neck.
Customers Who Bought This Also Bought