Order Entry
ContactUsLinkComponent
5193 ADAPTER, Vacuum Take-off, Short Stem
Adapter, Vacuum Take-Off, Short Stem, Ace Glass Incorporated
# de catalogue 89051-376
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
5193 ADAPTER, Vacuum Take-off, Short Stem
Adapter, Vacuum Take-Off, Short Stem, Ace Glass Incorporated
# de catalogue 89051-376
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise

Spécifications

  • Description:
    Vacuum Take-Off, Short Stem Adapter
  • Cat. no.:
    89051-376
  • Bottom Inner [ST] Joint Size:
    24/40
  • Supplier No.:
    5193-08

Spécifications

A propos de cet article

This vacuum take-off adapter features a short stem.

  • Stem length below joint of 30 mm
  • With hose connection
  • Fabricated from borosilicate glass

The stem length below the joint is 30mm. Use with a 3/8-inch or 5/16-inch I.D. tubing, size C hose connection.

Customers Who Bought This Also Bought