ContactUsLinkComponent
INDICATOR STRIP
pH Indicator Strips, pH-Fix, Macherey-Nagel
Catalog # 89049-924
Supplier:  Macherey-Nagel
Small Business Enterprise
pH Indicator Strips, pH-Fix, Macherey-Nagel
Catalog # 89049-924
Supplier:  Macherey-Nagel
Small Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    pH-Fix pH Test Strips, 6×85 mm
  • Cat. no.:
    89049-924
  • pH Range:
    0.0 - 14.0
  • pH Graduation:
    1.0
  • No. of Strips:
    100
  • Supplier No.:
    92110
Specifications
About this item
pH-Fix test strips allow fast pH testing directly at the point of interest.
  • Ready to use
  • Easy and safe
  • Packed in classic flat box
  • Several test pads for exact results
  • Safe analysis by long plastic handle
  • Fixed indicator dye does not leach into sample
Even in weakly buffered solutions, a safe and reliable pH determination becomes possible, as the strips can be immersed for a long time until the final reaction color has developed
pH-Fix test strips are tested and approved for medical applications and carry a CE-mark for in-vitro-diagnostics 98/79/EG or medical products 93/42/EWG. They meet the special demands of health care professionals and ensure safe results for medical pH-testing.
The colors of the individual test pads don't bleed and mix, which assures exact comparison to the color scale.
Certifications: ISO 13485 certified.
Customers Who Bought This Also Bought