ContactUsLinkComponent
pH Indicator Papers, Duotest, Machery Nagel
Catalogue # 89049-914
Fournisseur:  Macherey-Nagel
Small Business Enterprise
pH Indicator Papers, Duotest, Machery Nagel
Catalogue # 89049-914
Fournisseur:  Macherey-Nagel
Supplier Number:  90304
Small Business Enterprise
Spécifications
  • Cat. no.:
    89049-914
  • pH Range:
    5.0 - 8.0
  • pH Graduation:
    0.3
  • Supplier No.:
    90304
Spécifications
À propos de cet article
Duotest indicator papers combine two different indicator zones on a single strip
The zones are separated by a hydrophobic barrier, which effectively prevents mixing of the reaction colors and increases the mechanical stability.
Frequently Bought Together