ContactUsLinkComponent
Ovation Pipette Tips
Disposable Tips for MLA and Ovation Pipettors, VistaLab
Catalog # 89049-042
Supplier:  VistaLab
Disposable Tips for MLA and Ovation Pipettors, VistaLab
Catalog # 89049-042
Supplier:  VistaLab
Specifications
  • Volume:
    10 ml
  • Packed:
    VistaBulk™
  • Cat. no.:
    89049-042
  • Tip Style:
    Standard
  • Supplier No.:
    4058-6000
Specifications
About this item
Disposable polypropylene tips for use with MLA or ovation pipettors.
  • Non-pyrogenic
  • Non-sterile or sterile, RNase/DNase-free
  • Filtered or non-filtered
  • Low retention available
  • Trace metal certified available
VistaLab tips are available in a full range of volumes from 2 μl to 10 ml for Macro Volume pipettors.
These disposable tips are an economical choice and are available in a wide range of sizes.
Customers Who Bought This Also Bought