ContactUsLinkComponent
Liquid Sensing Tips
Axygen® Automation Tips for Tecan®Corning
Catalog # 89040-074
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
Axygen® Automation Tips for Tecan®Corning
Catalog # 89040-074
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
Specifications
  • Volume:
    20 µl
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Packed:
    96/Rack, 24 Racks/Case
  • Cat. no.:
    89040-074
  • Supplier No.:
    TT-20-CBK-HTR
Specifications
About this item
PP, with or without filter, sterile or non sterile.
  • Black tips are conductive for liquid level-sensing protocols
  • Optimised fine point for accurate dispensing
  • Certified RNase-, DNase- and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
EVO-style tips are compatible with Tecan Freedom EVO®, Caliper Zephyr® SPE, and BioTek® Precision™ robotic pipetting systems.
Genesis-style tips are compatible with Tecan Genesis Freedom, Freedom EVO, and MiniPrep robotic workstations, as well as Cavro®, PerkinElmer/Packard Multiprobe II HT, and Multiprobe II HT EX workstations.
Packaging: Packaged in plastic blister packs.
Customers Who Bought This Also Bought