ContactUsLinkComponent
Septum Stopper, Sleeve Type
Septum Stoppers, Sleeve Type, Chemglass
# de catalogue 89029-092
Fournisseur:  Chemglass
Small Business Enterprise
Septum Stopper, Sleeve Type
Septum Stoppers, Sleeve Type, Chemglass
# de catalogue 89029-092
Fournisseur:  Chemglass
Small Business Enterprise
Spécifications
  • Color:
    White
  • Description:
    Stopper, Sleeve Type, for 19/22 to 19/38 Outer Joints
  • Cat. no.:
    89029-092
  • Supplier No.:
    CG-3022-91
Spécifications
A propos de cet article
Sleeve type stoppers with hollow plugs and sleeve extensions.
  • Available in red or white rubber
The sleeve extensions fold down over the neck of a vessel and tubing. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.
Customers Who Bought This Also Bought