Septum Stoppers, Sleeve Type, Chemglass
# de catalogue 89029-092
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Color:White
- Description:Stopper, Sleeve Type, for 19/22 to 19/38 Outer Joints
- Cat. no.:89029-092
- Supplier No.:CG-3022-91
Spécifications
A propos de cet article
Sleeve type stoppers with hollow plugs and sleeve extensions.
- Available in red or white rubber
The sleeve extensions fold down over the neck of a vessel and tubing. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.
Customers Who Bought This Also Bought