ContactUsLinkComponent
PATCHXPRESS 50UL STERILE TIP
Automation Pipette Tips for PatchXpress Workstations, Thermo Scientific
# de catalogue 89025-942
Fournisseur:  Thermo Scientific
Automation Pipette Tips for PatchXpress Workstations, Thermo Scientific
# de catalogue 89025-942
Fournisseur:  Thermo Scientific
Spécifications
  • Volume:
    50 µl
  • Tip Color:
    Clear
  • Packed:
    96 Tips/Tray, 10 Trays/case
  • Sterile:
    +
  • For Use With:
    PatchXpress automated workstations
  • Supplier No.:
    902-261XP
Spécifications
A propos de cet article
Thermo Scientific provides a solution for your PatchXpress robotic pipette tips.
Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Each lot must pass a 15-point QC inspection before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
Customers Who Bought This Also Bought