Clean Room Adhesive Mats, Wearwell®
# de catalogue 89021-748
Fournisseur: Wearwell
Spécifications
- Color:Blue
- Description:Cleanroom anti-fatigue tacky mats, polyethylene
- Cat. no.:89021-748
- Packaging:30 sheets/pad
- Length:91 cm (3 ft.)
- Width:46 cm (18")
- Supplier No.:095.18X36BL
Spécifications
A propos de cet article
Effective and easy-to-use PE mat offers economical dust and dirt control for low-profile carpet, tile, or concrete surfaces.
- Economical, easy to use, effective dust and dirt control for low-profile carpet, tile, or concrete surfaces
- Tacky surface removes impurities from shoe soles before they can contaminate a cleanroom or anywhere dirt contamination is a hazard
- 30 layers of tough film laminated together in a stack
Once the top layer is contaminated, simply pull it off and discard.
Customers Who Bought This Also Bought