Spécifications
- Description:Isoflurane, pin-index bottle fill adapter
- Cat. no.:89012-522
- Supplier No.:911117
Spécifications
A propos de cet article
Precision vaporizers deliver specific anesthetic agents by percentage concentration.
- With automatic flowrate and temperature compensation
- Easy-to-read dial and sight glass for at-a-glance liquid level monitoring
- Integrated safety lock to prevent unintentional engagement
- Reusable and can be recapped and left on the bottle
Pin-indexed vaporizers require a pin-index bottle fill adapter to eliminate spillage, room pollution, and cross-agent contamination. The anti-spill bottle-top adapters allow the filling of a funnel-fill vaporizer without excessive spillage or pollution.