ContactUsLinkComponent
Cap
Vaporizers, VetEquip®
# de catalogue 89012-522
Fournisseur:  Vetequip
Vaporizers, VetEquip®
# de catalogue 89012-522
Fournisseur:  Vetequip
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Isoflurane, pin-index bottle fill adapter
  • Cat. no.:
    89012-522
  • Supplier No.:
    911117
Spécifications
A propos de cet article
Precision vaporizers deliver specific anesthetic agents by percentage concentration.
  • With automatic flowrate and temperature compensation
  • Easy-to-read dial and sight glass for at-a-glance liquid level monitoring
  • Integrated safety lock to prevent unintentional engagement
  • Reusable and can be recapped and left on the bottle
Pin-indexed vaporizers require a pin-index bottle fill adapter to eliminate spillage, room pollution, and cross-agent contamination. The anti-spill bottle-top adapters allow the filling of a funnel-fill vaporizer without excessive spillage or pollution.
Ordering information: Vaporizers include a manufacturer’s one-year money-back guarantee and seven-year replacement warranty.
Customers Who Bought This Also Bought