Spécifications
- Description:O2 regulator for E-cylinders
- Cat. no.:89012-466
- Supplier No.:901305
Spécifications
A propos de cet article
Carrier gas can be supplied from a small cylinder or large tank in the procedure room, on an anesthesia cart, or piped through a wall outlet.
- Gas-specific regulators have a DISS male connection and are preset to 50 psi
- Flowmeters can be ordered to replace existing units or added to create a custom machine with varying carrier gas combinations
Regulators and flowmeters include a manufacturer’s one-year money-back guarantee and seven-year replacement warranty.