ContactUsLinkComponent
Cap
Gas Supply Regulators and Flowmeters, VetEquip®
# de catalogue 89012-466
Fournisseur:  Vetequip
Cap
Gas Supply Regulators and Flowmeters, VetEquip®
# de catalogue 89012-466
Fournisseur:  Vetequip
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    O2 regulator for E-cylinders
  • Cat. no.:
    89012-466
  • Supplier No.:
    901305
Spécifications
A propos de cet article
Carrier gas can be supplied from a small cylinder or large tank in the procedure room, on an anesthesia cart, or piped through a wall outlet.
  • Gas-specific regulators have a DISS male connection and are preset to 50 psi
  • Flowmeters can be ordered to replace existing units or added to create a custom machine with varying carrier gas combinations
Regulators and flowmeters include a manufacturer’s one-year money-back guarantee and seven-year replacement warranty.
Customers Who Bought This Also Bought