ContactUsLinkComponent
VWR® Glove Box Holders, Epoxy-Coated
Catalogue # 89011-728
Fournisseur:  VWR International
undefined
VWR® Glove Box Holders, Epoxy-Coated
Catalogue # 89011-728
Fournisseur:  VWR International
Spécifications
  • Capacity:
    3 Boxes
  • Cat. no.:
    89011-728
  • W×H×D:
    9.6×25×43.2 cm (3¾×9¹³⁄₁₆×17")
Spécifications
À propos de cet article
Epoxy-coated steel wire racks hold most major brands of glove boxes.
  • Steel wire racks hold most major glove box brands
Choose from a single-box holder or a three-box holder.
Ordering information: Supplied with wall-mounting screws.
Frequently Bought Together