Blood Cell Counting Vial, Globe Scientific
# de catalogue 89009-318
Fournisseur: Globe Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Cap included:Yes
- Cap Type:Attached snap cap
- Description:Blood cell counting vial
- Packaging:250/Bag, 4 Bags/Case
- Supplier No.:107047
Spécifications
A propos de cet article
These cell counter vials come with a polyethylene snap-on lid.
The vials are graduated at the 10 and 20 ml marks.
Customers Who Bought This Also Bought