ContactUsLinkComponent
DILU-VIALS 20ML CS1000
Blood Cell Counting Vial, Globe Scientific
# de catalogue 89009-318
Fournisseur:  Globe Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
DILU-VIALS 20ML CS1000
Blood Cell Counting Vial, Globe Scientific
# de catalogue 89009-318
Fournisseur:  Globe Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
  • Cap included:
    Yes
  • Cap Type:
    Attached snap cap
  • Description:
    Blood cell counting vial
  • Packaging:
    250/Bag, 4 Bags/Case
  • Supplier No.:
    107047
Spécifications
A propos de cet article
These cell counter vials come with a polyethylene snap-on lid.
The vials are graduated at the 10 and 20 ml marks.
Customers Who Bought This Also Bought