ContactUsLinkComponent
VWR* Bag
VWR® Reclosable UV-Protectant Amber Bags
# de catalogue 89005-328
Fournisseur:  VWR International
VWR* Bag
VWR® Reclosable UV-Protectant Amber Bags
# de catalogue 89005-328
Fournisseur:  VWR International
Spécifications
  • W×L:
    203×203 mm (8×8")
  • Cat. no.:
    89005-328
Spécifications
A propos de cet article
Ideal for storage and transport of light-sensitive products or medications.
  • Re-closable design
  • Amber tint protects against UV radiation
  • Easy to identify with amber tint
Re-closable 3 mil bags feature a wide, white zipper to allow for repeated use and provides protection against dust and moisture. Amber tint makes bags easily identifiable.
Certifications: Bags meet light transmission rates between the ranges of full shade at DIN 53775 and DIN 53387, as well as reduced shade at DIN 53375.
Customers Who Bought This Also Bought