Order Entry
ContactUsLinkComponent
VWR* Bag
VWR® Reclosable Clear Bags
# de catalogue 89005-308
Fournisseur:  VWR International
VWR* Bag
VWR® Reclosable Clear Bags
# de catalogue 89005-308
Fournisseur:  VWR International

Spécifications

  • W×L:
    203×254 mm (8×10")
  • Cat. no.:
    89005-308

Spécifications

A propos de cet article

Standard clear re-closable bags are available in 2 and 4 mm thickness.

  • Clear siding for quick visual identification of contents
  • Wide zipper eases loading
  • Re-closable design

A wide zipper eases loading and provides a strong, secure seal. Bags 15.2 cm (6") in height or smaller are packed in perforated dispenser bags; all other bags are packed in inner bags without the dispenser bag.

Customers Who Bought This Also Bought