Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Description:E. cloni 10G Chemically Competent Cells (≥1 x 10^9 cfu/µg DNA)
- Genotypes:E. cloni 10G: F- mcrA Δ(mrr-hsdRMS-mcrBC) endA1 recA1 Φ80dlacZΔM15 ΔlacX74 araD139 Δ(ara,leu)7697 galU galK rpsL nupG λ- tonA.
- Cat. no.:89002-676
- Packaging:Duo
- No. of Reactions:24
- Supplier No.:60107-2
Spécifications
À propos de cet article
Lucigen’s E
Lucigen offers the following choices of strain: 1) E. cloni 10G Competent Cells: Routine cloning, subcloning, and plasmid isolation with or without blue/white screening. 2) E. cloni 10GF′ Competent Cells: Contain the F′ plasmid for infection with M13 to produce ssDNA.
And the following choices of efficiency: 1) E. cloni 10G Chemically Competent Cells (> 1 × 10^9 cfu/µg pUC DNA; (> 1 × 10^8 in 96-well plates). Highly efficient competent cells for routine cloning applications. Available in one transformation per tube (SOLOs) or two transformations per tube (DUOs). Also available in 96-well plates (which can be divided into 24-well seqments) for higher throughput. 2) E. cloni 10GF´ Chemically Competent Cells (≥5 × 10^8 cfu/µg pUC DNA). An excellent choice for producing single-stranded DNA for sequencing, mutagenesis, etc. 3) E. cloni 10G Chemically Competent Cells, Subcloning Grade (≥1 × 10^6 cfu/µg pUC DNA). The best value available anywhere for simple cloning and plasmid propagation.
cloni competent cells share the most useful genetic elements of standard cloning strains like DH5α™, DH10B™, JM109, TOP10, etc. and directly replace them in cloning protocols. E. cloni chemically competent cells provide the performance researchers need with ease of use-only common laboratory equipment is required. These cells provide solutions for a wide range of applications at unparalled economical prices.