Order Entry
ContactUsLinkComponent
KIMAX* Heavy-Duty Centrifuge Tube
KIMAX® Centrifuge Tubes, Graduated, Conical-Bottom, with Screw Caps, Kimble Chase
# de catalogue 89002-216
KIMAX* Heavy-Duty Centrifuge Tube
KIMAX® Centrifuge Tubes, Graduated, Conical-Bottom, with Screw Caps, Kimble Chase
# de catalogue 89002-216

Spécifications

  • Cap Size:
    24-410
  • Volume:
    40 ml
  • Description:
    Heavy-Duty Tube
  • Maximum RCF:
    2980×g
  • O.D.×L:
    29×140 mm
  • Supplier No.:
    45200-40

Spécifications

A propos de cet article

Tubes are graduated and calibrated to contain.

  • Centrifuge tubes are made of borosilicate glass

Scale, legend, and marking spot are printed in durable white ceramic enamel. Tubes have a screw-thread finish and include an autoclavable phenolic cap with PTFE-faced rubber liner. Heavy-duty tubes are made with heavier walled tubing to withstand higher centrifugation speeds.

Customers Who Bought This Also Bought