ContactUsLinkComponent
KIMAX* Glass Beads
KIMAX® Solid Borosilicate Glass Beads, Kimble Chase
# de catalogue 89001-520
KIMAX* Glass Beads
KIMAX® Solid Borosilicate Glass Beads, Kimble Chase
# de catalogue 89001-520
Spécifications
  • Ø:
    6 mm
  • Cat. no.:
    89001-520
  • No. of Beads:
    1800/container; 75/cu.in.
  • Supplier No.:
    13500-6
Spécifications
A propos de cet article
Highly resistant to attack by most cold liquids, hot liquids, and vapors
Suitable for use as mixing beads, boiling stones, or packing for distillation columns. Beads are durable and will not disintegrate or affect delicate compounds.
Ordering information: Packed in 0.45kg (1lb.) containers with a total volume of 360cm3 (22 cu. in.).
Customers Who Bought This Also Bought