Phenolic Screw Thread Closure, Kimble Chase
# de catalogue 89001-504
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
Spécifications
- Cap Size:15-415
- Height:16 mm
- Cat. no.:89001-504
- Supplier No.:45066C-15
Spécifications
A propos de cet article
Caps have mineral-filled phenolic material to withstand the effects of repeated autoclaving.
Rubber-lined caps retain adhesive properties and meet ASTM Specification E982, Class A requirements. PTFE-faced rubber-lined caps meet ASTM specification E982, Class B requirements.
Unlined closures have excellent chemical resistance and are autoclavable.
Customers Who Bought This Also Bought