ContactUsLinkComponent
Phenolic PTFE-Faced Screw Cap
Phenolic Screw Thread Closure, Kimble Chase
# de catalogue 89001-504
Phenolic PTFE-Faced Screw Cap
Phenolic Screw Thread Closure, Kimble Chase
# de catalogue 89001-504
Spécifications
  • Cap Size:
    15-415
  • Height:
    16 mm
  • Cat. no.:
    89001-504
  • Supplier No.:
    45066C-15
Spécifications
A propos de cet article
Caps have mineral-filled phenolic material to withstand the effects of repeated autoclaving.
Rubber-lined caps retain adhesive properties and meet ASTM Specification E982, Class A requirements. PTFE-faced rubber-lined caps meet ASTM specification E982, Class B requirements.
Unlined closures have excellent chemical resistance and are autoclavable.
Customers Who Bought This Also Bought