Phenolic Screw Thread Closure, Kimble Chase
# de catalogue 89001-492
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Cap Size:13-415
- Color:Black
- Height:14 mm
- Cat. no.:89001-492
- Supplier No.:45066B-13
Spécifications
A propos de cet article
Caps have mineral-filled phenolic material to withstand the effects of repeated autoclaving.
Rubber-lined caps retain adhesive properties and meet ASTM Specification E982, Class A requirements. PTFE-faced rubber-lined caps meet ASTM specification E982, Class B requirements.
Unlined closures have excellent chemical resistance and are autoclavable.
Customers Who Bought This Also Bought