Order Entry
ContactUsLinkComponent
VWR Signature* Pipettor, Fixed Volume
VWR® Signature™ Single Channel Pipettors, Fixed Volume
Catalog # 83009-758
Supplier:  VWR International
undefined
VWR® Signature™ Single Channel Pipettors, Fixed Volume
Catalog # 83009-758
Supplier:  VWR International

Specifications

  • Volume:
    5 µl
  • Cat. no.:
    83009-758
  • For Use With:
    VWR UNIV TIPS
  • Accuracy:
    ±1.6%
  • Precision:
    ±1%

Specifications

About this item

Ideal for applications where specific volumes are used repeatedly, these slender, contoured pipettor bodies fit comfortably in either hand, as well as in large or small hands.

  • Ten fixed volumes available
  • Narrow shaft for universal tip fit
  • Removable, adjustable tip ejector
  • Fully autoclavable
  • VWR® two-year limited warranty
  • Pipettor stands hold six pipettors

In addition, maintenance-free seals and highly polished pistons provide exceptional accuracy and precision, as well as allow the pipettors to be autoclaved repeatedly without recalibration

The corrosion-resistant, stainless steel tip ejector is removable for pipetting into narrow tubes and it can be adjusted to accommodate various types of tips. The slender, tapered shaft provides universal tip compatibility and is highly chemical-resistant.

Frequently Bought Together