Polypropylene Screw Caps, BrandTech
# de catalogue 83009-034
Fournisseur: Brandtech
Spécifications
- Cap Size:GL32
- Cat. no.:83009-034
- Supplier No.:V83312
Spécifications
A propos de cet article
Translucent screw caps are for use with BrandTech polypropylene and low-density polyethylene bottles.
- Autoclavable translucent polypropylene
- Fits VITLAB and many other bottles with standard thread
Packaging: Bulk packed.
Customers Who Bought This Also Bought