VITLAB® Graduated Pitchers, Polypropylene, BrandTech
# de catalogue 83008-778
Fournisseur: Brandtech
Spécifications
- Capacity:3 L (101.4 oz.)
- Graduation Interval:50 ml
- Cat. no.:83008-778
- Supplier No.:V484941
Spécifications
A propos de cet article
Made of durable, translucent polypropylene.
- Pitchers with molded graduations areavailable with or without overprint
- Nesting pitchers have screened overprint
- Blue printing provides high contrast with many liquids
Standard graduated pitchers have molded graduations with or without a blue overprint for high legibility. Nesting pitchers conserve bench and storage space and have blue screened graduations for easy, accurate measurements.
Customers Who Bought This Also Bought