SMS Coveralls, International ENVIROGUARD™
# de catalogue 83007-184
Fournisseur: International Enviroguard
Spécifications
- Clothing Size:XXL
- Garment application:Laboratory
- Color:Denim Blue
- Cat. no.:83007-184
- Supplier No.:2229BXXL
Spécifications
A propos de cet article
Coveralls are constructed of three-layer, spunbonded-meltblown-spunbonded material, which offers splash resistance yet is cool and comfortable to wear
Garments may be used for pharmaceutical and general industrial applications, as well as manufacturing, asbestos removal, and assembly operations. Coveralls feature a zipper front, hood, boots, and elastic wrists, ankles, and back. Fabric weight: 60g.
Customers Who Bought This Also Bought