96-Well No-Bottom Plates, Greiner Bio-One
Catalogue # 82051-528
Fournisseur: Greiner Bio-One
Product available on GSA Advantage®
Spécifications
- Color:Black
- Cat. no.:82051-528
- Supplier No.:655000-06
Spécifications
À propos de cet article
Plates are made from virgin polystyrene resin and molded without the bottom of the plate.
- Virgin polystyrene construction
- Ideal for work with nonpolar proteins and peptides
- Medium binding surface
- Designed without a bottom
They feature chimney-well design to help prevent cross-contamination
Half-area plates maintain ANSI/SBS standards and well-to-well pitch, yet feature a well diameter design that offers one-half the well surface area of standard plates. This format allows a 50% sample volume reduction without moving to high well density plates. Half-area plates feature a medium binding polystyrene surface, which is ideal for work with nonpolar proteins and peptides.
Available in standard or half-area formats.
Frequently Bought Together