ContactUsLinkComponent
384-Well Plate
384-Well Microplates, Polypropylene, Greiner Bio-One
# de catalogue 82051-328
Fournisseur:  Greiner Bio-One
Product available on GSA Advantage®
384-Well Plate
384-Well Microplates, Polypropylene, Greiner Bio-One
# de catalogue 82051-328
Fournisseur:  Greiner Bio-One
Product available on GSA Advantage®
Spécifications
  • Sterility:
    Non sterile
  • Color:
    Clear
  • Working Volume:
    13 - 120 µl
  • Supplier No.:
    781280
Spécifications
A propos de cet article
These 384-well microplates are manufactured from ClearChoice™ virgin polypropylene resin and produced with uniform external dimensions and tolerances to meet ANSI recommendations for compatibility with equipment and robotics.
  • Nonpyrogenic
  • High temperature tolerance
  • Chemically resistant
  • Free of RNase, DNase, and human DNA
  • Stackable
  • Autoclavable
Microplates are biologically inert, exhibit low binding characteristics, offer a high temperature tolerance, and are resistant to many standard laboratory chemicals
Microplates feature a square-round top well geometry to minimize wicking and air bubble formation. Flat-bottom plates or conical-bottom plates are available to enable complete sample recovery, and raised well rims facilitate thermal, adhesive, or cap mat sealing. Stackable and autoclavable. The well-to-well spacing is 4.5mm.
Free of detectable RNase, DNase, human DNA, and pyrogens. All plates have a footprint conforming to the ANSI/SBS 1-2004 Standard.
These properties provide optimal conditions for laboratory specimen and compound storage.
Customers Who Bought This Also Bought