Spécifications
- Sterility:Non sterile
- Color:Clear
- Working Volume:13 - 120 µl
- Supplier No.:781280
Spécifications
A propos de cet article
These 384-well microplates are manufactured from ClearChoice™ virgin polypropylene resin and produced with uniform external dimensions and tolerances to meet ANSI recommendations for compatibility with equipment and robotics.
- Nonpyrogenic
- High temperature tolerance
- Chemically resistant
- Free of RNase, DNase, and human DNA
- Stackable
- Autoclavable
Microplates are biologically inert, exhibit low binding characteristics, offer a high temperature tolerance, and are resistant to many standard laboratory chemicals
Microplates feature a square-round top well geometry to minimize wicking and air bubble formation. Flat-bottom plates or conical-bottom plates are available to enable complete sample recovery, and raised well rims facilitate thermal, adhesive, or cap mat sealing. Stackable and autoclavable. The well-to-well spacing is 4.5mm.
Free of detectable RNase, DNase, human DNA, and pyrogens. All plates have a footprint conforming to the ANSI/SBS 1-2004 Standard.
These properties provide optimal conditions for laboratory specimen and compound storage.