Spécifications
- Format:LUMITRAC™ 600
- Sterility:Non sterile
- Color:White
- Surface Treatment:High binding
- Working Well Volume:10 - 130 µl
- Maximum Well Volume:138 µL
- Supplier No.:781074
Spécifications
A propos de cet article
Microplates are manufactured from virgin polystyrene resin and are designed to meet industry standard dimension specifications to facilitate compatibility with microplate handling and robotics.
- Virgin polystyrene construction
- Free of RNase, DNase, human DNA, and pyrogens
- Designed to reduce air bubble formation in wells
- Stackable
All plates feature a flat bottom and a rounded square top (square with round corners) designed to minimize wicking and reduce air bubble formation, while increasing volume capacity and surface area
Plates are free of detectable RNase, DNase, human DNA, and pyrogens. All plates have a footprint conforming to the ANSI/SBS 1-2004 Standard.
White LUMITRAC™ plates are for luminescence assays and black FLUOTRAC™ plates are for fluorescence assays. Clear-bottom plates are manufactured with µClear™ mircro-thin polystyrene film, and are suggested for assays designed for luminescence and fluorescence detection in which imaging from the bottom is required. Plates are stackable and supplied without lids.