Inoculating Loops and Needles, Sterile, Greiner Bio-One
# de catalogue 82051-138
Fournisseur: Greiner Bio-One
Product available on GSA Advantage®
Spécifications
- Length:20 cm (77/8")
- Color:White
- Description:1 µL Inoculating Loops
- Cat. no.:82051-138
- Packaging:Individually Packed
- Supplier No.:731161
Spécifications
A propos de cet article
Inoculating loops allow easier bacterial collection and inoculation.
- Color-coded
- Polystyrene construction
- Sterile
Inoculating needles are ideal for picking up single colonies.
Customers Who Bought This Also Bought