ContactUsLinkComponent
CELLSTAR® Cell Culture Multiwell Plates, Polystyrene, Greiner Bio-One
Catalogue # 82050-844
Fournisseur:  Greiner Bio-One
Product available on GSA Advantage®
undefined
CELLSTAR® Cell Culture Multiwell Plates, Polystyrene, Greiner Bio-One
Catalogue # 82050-844
Fournisseur:  Greiner Bio-One
Supplier Number:  657165
Product available on GSA Advantage®
Spécifications
  • No. of wells:
    6
  • Cat. no.:
    82050-844
  • Packaging:
    5/Sleeve, 24 Sleeves/Case
  • Supplier No.:
    657165
Spécifications
À propos de cet article
Cell Culture Multiwell Plates allow for optimal cultivation conditions for sensitive cells and cells cultivated under restricted growth conditions.
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Virgin polystyrene construction
  • Chimney well design helps reduce cross-contamination
Surface treatments provide the conditions for improved cell adherence, accelerated cell expansion, and they increase overall cell yield.
Manufactured from optically clear virgin polystyrene, these flat-bottom plates are supplied sterile and pyrogen-free with a printed lot number for traceability. The chimney well design and low evaporation lids with condensation rings reduce the risk of cross-contamination. Plates are available tissue culture treated for the culture of anchorage-dependent cells or nontreated for suspension cell culture applications. Excellent heat exchange within each plate is ensured during incubation through air circulation spaces between stacked plates. Uniform design makes them suitable for manual and automated handling. The well depth and diameter also allows the use of specific tissue culture inserts.
Sterilized by radiation. DNase-, RNase-, and human DNA-free. Nonpyrogenic and noncytotoxic.
Note: The nontreated, 24-well plates packaged 12 per sleeve are black, nonsterile, and supplied without lid.
Ordering information: Supplied in sterile and user friendly packaging.
Frequently Bought Together