96-Well Black and White Microplates, Polystyrene, Greiner Bio-One
Product Details & Documents
Black and white microplates are manufactured from virgin polystyrene resin and are designed to meet industry standard dimension specifications to facilitate compatibility with microplate handling and robotics.
- Free of RNase, DNase, and human DNA
- Nonpyrogenic
- Virgin polystyrene construction
- Solid bottoms or clear bottoms
All of the black and white plates have square chimney wells with flat bottoms
LUMITRAC™ and FLUOTRAC™ formats are offered to allow a wide variety of applications, including cell-based assays. White LUMITRAC™ plates are for luminescence assays and black FLUOTRAC™ plates are for fluorescence assays.
Plates are available with solid or clear bottoms. Clear bottom plates are manufactured with µClear™ micro-thin polystyrene film, and are suggested for assays designed for luminescence and fluorescence detection in which imaging from the bottom is required. Plates are supplied without lids.
These 96-well polystyrene microplates plates are free of detectable RNase, DNase, human DNA, and pyrogens. All plates have a footprint conforming to the ANSI/SBS 1-2004 Standard.
Spécifications
- Sterility:Non sterile
- Color:Black
- Surface Treatment:Medium binding
- Working Well Volume:382 µl
- Maximum Well Volume:25–340 µL